Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1922

Sigma-Aldrich

K67

≥98% (HPLC)

Synonym(s):

2-Acetonyl-1,4-bis[(4-ethoxybenzensulfonyl)amino]naphthalene, N,N′-(2-(2-Oxopropyl)naphthalene-1,4-diyl)bis(4-ethoxybenzenesulfonamide), N-[2-Acetonyl-4-(4-ethoxybenzenesulfonylamino)naphthalene-1-yl]-4-ethoxybenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H30N2O7S2
CAS Number:
Molecular Weight:
582.69
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(CC1=C(NS(C2=CC=C(OCC)C=C2)(=O)=O)C3=C(C=CC=C3)C(NS(C4=CC=C(OCC)C=C4)(=O)=O)=C1)=O

InChI key

VUIVGOOWLHGDPZ-UHFFFAOYSA-N

Biochem/physiol Actions

K67 is a specific inhibitor against the interaction between KEAP1 (Kelch-like ECH-associated protein 1) DC (double glycine repeat and C-terminal region) domain and S349-phosphorylated (human Ser349, mouse Ser351) KIR (Keap1-interacting region) of p62/SQSTM1, thereby preventing phospho-p62 from blocking KEAP1-DC and NRF2 (nuclear factor erythroid 2-related factor 2) DLGex motif association. K67 effectively inhibits the proliferation of HCC (hepatocellular carcinoma) cultures (by 59% of Huh7 cells post 72 h 50 μM K67 treatment) with high cellular p62 S351-phosphorylation by restoring KEAP1-driven NRF2 ubiquitination and degradation.
K67 is a specific inhibitor against the interaction between KEAP1 (Kelch-like ECH-associated protein 1) DC (double glycine repeat and C-terminal region) domain and S349-phosphorylated (human Ser349, mouse Ser351) KIR (Keap1-interacting region) of p62/SQSTM1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Yasuda et al.
Bioorganic & medicinal chemistry letters, 26(24), 5956-5959 (2016-11-15)
The Keap1-Nrf2 system is involved not only in biological defense but also in malignancy progression and chemoresistance. The ubiquitin-binding protein p62/Sqstm1 (p62), which is highly expressed in several cancers, competes with Nrf2 for Keap1 binding, leading to activation of Nrf2-mediated
Tetsuya Saito et al.
Nature communications, 7, 12030-12030 (2016-06-28)
p62/Sqstm1 is a multifunctional protein involved in cell survival, growth and death, that is degraded by autophagy. Amplification of the p62/Sqstm1 gene, and aberrant accumulation and phosphorylation of p62/Sqstm1, have been implicated in tumour development. Herein, we reveal the molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service