Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05987

Sigma-Aldrich

Keap1-Nrf2 Interaction Probe, ML334

Synonym(s):

Keap1-Nrf2 Interaction Probe, ML334, (1S,2R)-2-((S)-1-((1,3-dioxoisoindolin-2-yl)methyl)-1,2,3,4-tetrahydroisoquinoline-2-carbonyl)cyclohexanecarboxylic acid, (SRS)-LH601, Nrf Activator VI

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$342.00

$342.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$342.00

About This Item

Empirical Formula (Hill Notation):
C26H26N2O5
CAS Number:
Molecular Weight:
446.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$342.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

cream

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

SMILES string

N4(C(=O)c5c(cccc5)C4=O)C[C@H]1N(CCc3c1cccc3)C(=O)[C@H]2[C@H](CCCC2)C(=O)O

InChI

1S/C26H26N2O5/c29-23(20-11-5-6-12-21(20)26(32)33)27-14-13-16-7-1-2-8-17(16)22(27)15-28-24(30)18-9-3-4-10-19(18)25(28)31/h1-4,7-10,20-22H,5-6,11-15H2,(H,32,33)/t20-,21+,22-/m1/s1

InChI key

LNENLABLFGGAFF-BHIFYINESA-N

General description

A cell-permeable tetrahydroisoquinoline derivative that acts as a direct, non-covalent inhibitor of Keap1 - Nrf2 (Kelch-like ECH-associated protein 1 - nuclear factor erythroid 2-related factor 2) protein - protein interaction (Kd = 1 µM) that causes the dissociation of Nrf2 from keap1 in the cytosol. Exhibits ARE-inducing activity (EC50 = 18 µM in U2OS Keap1-Nrf2 functional assay) and promotes Nrf2 translocation to the nucleus (EC50 = 12 µM). Has good solubility in aqueous buffers (>100 µM in phosphate buffered saline).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
Keap1–Nrf2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hu, L., et al, 2013, Bioorg. Med. Chem. Lett.23, 3039.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service