Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1902

Sigma-Aldrich

ISAM-140

≥98% (HPLC)

Synonym(s):

(±)-Isopropyl 4-(Furan-2-yl)-2-methyl-1,4-dihydrobenzo-[4,5]imidazo[1,2-a]pyrimidine-3-carboxylate, 4-(2-Furanyl)-4,10-dihydro-2-methyl-pyrimido[1,2-a]benzimidazole-3-carboxylic acid 1-methylethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19N3O3
CAS Number:
Molecular Weight:
337.37
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1=C(C(OC(C)C)=O)C(C2=CC=CO2)N3C(N1)=NC4=C3C=CC=C4

Biochem/physiol Actions

ISAM-140 is a potent and selective A2B adenosine receptor antagonist (A2BAR). ISAM-140 exhibits remarkable selectivity over A1, A2A, and A3 adenosine receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdelaziz El Maatougui et al.
Journal of medicinal chemistry, 59(5), 1967-1983 (2016-01-30)
Three novel families of A2B adenosine receptor antagonists were identified in the context of the structural exploration of the 3,4-dihydropyrimidin-2(1H)-one chemotype. The most appealing series contain imidazole, 1,2,4-triazole, or benzimidazole rings fused to the 2,3-positions of the parent diazinone core.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service