Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1638

Sigma-Aldrich

VX-745

≥98% (HPLC)

Synonym(s):

5-(2,6-Dichlorophenyl)-2-(2,4-difluorophenylthio)-6H-pyrimido[1,6-b]pyridazin-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H9Cl2F2N3OS
CAS Number:
Molecular Weight:
436.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

FC1=C(SC2=NN(C=N3)C(C=C2)=C(C4=C(Cl)C=CC=C4Cl)C3=O)C=CC(F)=C1

InChI

1S/C19H9Cl2F2N3OS/c20-11-2-1-3-12(21)17(11)18-14-5-7-16(25-26(14)9-24-19(18)27)28-15-6-4-10(22)8-13(15)23/h1-9H

InChI key

VEPKQEUBKLEPRA-UHFFFAOYSA-N

Gene Information

human ... MAPK14(1432)

Biochem/physiol Actions

VX-745 is a selective blood-brain barrier penetrant inhibitor of p38 MAPKα. VX-745 showed 20-fold selectivity for p38α over p38β and a Ki value of 220 nM. VX-745 has been shown to reduce inflammation in an arthritic (AIA) mouse model and to improve performance in aged rats.

Other Notes

VX-745 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the VX-745 probe summary on the Chemical Probes Portal website.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaodong Chen et al.
Journal of cancer research and clinical oncology, 145(6), 1471-1484 (2019-04-26)
To study an association between IL-6 signaling and resistance to radiotherapy of prostate cancer (PCa) and explore the molecular mechanisms involved. IL-6 expressing C4-2 and CWR22Rv1 (C4-2IL-6/CWRIL-6) and vector control (C4-2vec/CWRvec) cell lines were developed. Radiation-sensitivities of these cells were
Mehdi Bouhaddou et al.
Cell, 182(3), 685-712 (2020-07-10)
The causative agent of the coronavirus disease 2019 (COVID-19) pandemic, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has infected millions and killed hundreds of thousands of people worldwide, highlighting an urgent need to develop antiviral therapies. Here we present a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service