Skip to Content
MilliporeSigma
All Photos(1)

Documents

L-050

Supelco

Lidocaine-D10 solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12D10N2O
CAS Number:
Molecular Weight:
244.40
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

Lidocaine is a local anesthetic and antiarrhythmic drug. Lidocaine is often added to cocaine as a diluent. A Certified Snap-N-Spike® Solution for use in clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research applications by LC/MS or GC/MS.

Application



  • HPLC-MS/MS measurement of lidocaine in rat skin and plasma: A study utilized HPLC-MS/MS to measure Lidocaine concentrations in rat skin and plasma, demonstrating the method′s effectiveness for monitoring drug delivery from medicated plasters. This research supports the use of Lidocaine-D₁₀ as an internal standard for pharmacokinetic and bioanalytical assays, particularly in studies concerning topical anesthesia and pain relief applications (Matteo et al., 2020).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service