Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1480

Sigma-Aldrich

Alclometasone dipropionate

≥98% (HPLC)

Synonym(s):

Alclometasone Dipropanoate, Sch 22219, (7α,11β,16α)-7-Chloro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 7α-Chloro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione 17,21-di(propionate), Alclometasone 17,21-dipropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37ClO7
CAS Number:
Molecular Weight:
521.04
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Cl[C@H]1[C@@H]2[C@@H]([C@@]4(C(=CC(=O)C=C4)C1)C)[C@H](C[C@]3([C@H]2C[C@H]([C@]3(OC(=O)CC)C(=O)COC(=O)CC)C)C)O

InChI

1S/C28H37ClO7/c1-6-22(33)35-14-21(32)28(36-23(34)7-2)15(3)10-18-24-19(29)12-16-11-17(30)8-9-26(16,4)25(24)20(31)13-27(18,28)5/h8-9,11,15,18-20,24-25,31H,6-7,10,12-14H2,1-5H3/t15-,18+,19-,20+,24-,25+,26+,27+,28+/m1/s1

InChI key

DJHCCTTVDRAMEH-DUUJBDRPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Alclometasone dipropionate is a glucocorticoid receptor agonist that mimics the metabolic, anti-inflammatory, immunosuppressive, antipruritic, and vasoconstrictive effects of natural glucocorticoids.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Aoyama et al.
Drug metabolism and disposition: the biological fate of chemicals, 18(4), 409-417 (1990-07-01)
The metabolic fate of alclometasone dipropionate (ADP or S-3460) has been studied in rats, rabbits, and mice, with consideration of their interspecies differences. Several reference compounds related to ADP were synthesized for the determination and identification of the metabolites. After
Y Nishibe et al.
Japanese journal of pharmacology, 50(4), 435-443 (1989-08-01)
The effect of successive administration of the corticosteroid alclometasone dipropionate (ACM) on the hepatic drug-metabolizing system was examined using male and female rats. Although some pharmacological changes such as increases in plasma enzyme activity, lipid level and protein concentration appeared

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service