Skip to Content
MilliporeSigma
Alclometasone dipropionate ≥98% (HPLC)

Alclometasone dipropionate

Alclometasone dipropionate ≥98% (HPLC)
Synonym(s):
(7α,11β,16α)-7-Chloro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 7α-Chloro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione 17,21-di(propionate), Alclometasone 17,21-dipropionate
Empirical Formula (Hill Notation):
C28H37ClO7
CAS Number:
Molecular Weight:
521.04
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2