Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1427

Sigma-Aldrich

AZ3146

≥98% (HPLC)

Synonym(s):

9-Cyclopentyl-7,9-dihydro-2-[[2-methoxy-4-[(1-methyl-4-piperidinyl)oxy]phenyl]amino]-7-methyl-8H-purin-8-one, AZ-3146

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32N6O3
CAS Number:
Molecular Weight:
452.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN1CCC(OC2=CC(OC)=C(NC3=NC=C4C(N(C5CCCC5)C(N4C)=O)=N3)C=C2)CC1

InChI

1S/C24H32N6O3/c1-28-12-10-17(11-13-28)33-18-8-9-19(21(14-18)32-3)26-23-25-15-20-22(27-23)30(24(31)29(20)2)16-6-4-5-7-16/h8-9,14-17H,4-7,10-13H2,1-3H3,(H,25,26,27)

InChI key

YUKWVHPTFRQHMF-UHFFFAOYSA-N

Biochem/physiol Actions

AZ3146 is a potent and specific inhibitor of monopolar spindle 1 kinase (Mps1; IC50 = 35 nM), an important regulator of Spindle Assembly Checkpoint (SAC). AZ3146 treated HeLa cells override SAC, and quickly enter mitosis, with a reduced mitosis completion time of 32 minutes. 50% of treated cells entered anaphase without complete chromosomal alignment.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federica Schiavoni et al.
Nature communications, 13(1), 1731-1731 (2022-04-03)
Aneuploidy results in decreased cellular fitness in many species and model systems. However, aneuploidy is commonly found in cancer cells and often correlates with aggressive growth, suggesting that the impact of aneuploidy on cellular fitness is context dependent. The BRG1
Cayetana Vázquez-Diez et al.
Current biology : CB, 29(5), 865-873 (2019-02-19)
Chromosome segregation errors during mammalian preimplantation development cause "mosaic" embryos comprising a mixture of euploid and aneuploid cells, which reduce the potential for a successful pregnancy [1-5], but why these errors are common is unknown. In most cells, chromosome segregation
Anand Vasudevan et al.
Developmental cell, 52(4), 413-428 (2020-02-26)
High levels of cancer aneuploidy are frequently associated with poor prognosis. To examine the relationship between aneuploidy and cancer progression, we analyzed a series of congenic cell lines that harbor single extra chromosomes. We found that across 13 different trisomic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service