Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0761

Sigma-Aldrich

Gossypin

≥90% (HPLC)

Synonym(s):

2-(3,4-Dihydroxyphenyl)-8-(β-D-glucopyranosyloxy)-3,5,7-trihydroxy- 4H-1-benzopyran-4-one, 3,3′,4′,5,7,8-Hexahydroxyflavone 8-glucoside, Gossypetin 8-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O13
CAS Number:
Molecular Weight:
480.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

Hibiscus vitifolius

Assay

≥90% (HPLC)

form

powder

storage condition

desiccated

color

, light yellow to dark green-yellow

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2c(O)cc(O)c3C(=O)C(O)=C(Oc23)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O13/c22-5-11-13(27)15(29)17(31)21(32-11)34-19-10(26)4-9(25)12-14(28)16(30)18(33-20(12)19)6-1-2-7(23)8(24)3-6/h1-4,11,13,15,17,21-27,29-31H,5H2/t11-,13-,15+,17-,21+/m1/s1

InChI key

SJRXVLUZMMDCNG-KKPQBLLMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gossypin has anti-inflammatory properties. It inhibits galactose-induced cataract formation and reduces β-amyloid-induced toxicity. Gossypin provides protection against carbon tetrachloride-induced toxicity and bis(2-choloroethyl) sulfide-induced dermal toxicity. It also induces apoptosis in myeloma cells.[1] Gossypin has an anticonvulsant property.[2]

Biochem/physiol Actions

Gossypin is an anti-inflammatory pentahydroxyflavone glucoside isolated from Hibiscus vitifolius that is used as an herbal remedy for diabetes, jaundice, and inflammation. Gossypin potently inhibits human cancer cell proliferation, including melanoma and glioma cells. Gossypin directly binds and inhibits BRAFV600E and CDK4 kinases, and also regulates Chk1 and Cdc25C in U251 glioma cells.
Gossypin is an anti-inflammatory pentahydroxyflavone glucoside that directly binds and inhibits BRAFV600E and CDK4 kinases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anticonvulsant activity of bioflavonoid gossypin
Bangladesh Journal of Pharmacology, 4(1), 51-54 (2013)
Gossypin as a novel selective dual inhibitor of V-RAF murine sarcoma viral oncogene homolog B1 and cyclin-dependent kinase 4 for melanoma
Bhaskaran S, et al.
Molecular Cancer Therapeutics (2013)
Kuei-Chuan Chan et al.
Journal of the science of food and agriculture, 96(2), 381-391 (2015-01-24)
Previous studies have shown that mulberry polyphenolic compounds have an anti-atherosclerotic effect in rabbits. Apoptosis of vascular smooth muscle cells (VSMCs) is the key determinant of the number of VSMCs in remodeling. To examine the effect of mulberry polyphenol extracts
Gema Núñez-López et al.
Enzyme and microbial technology, 122, 19-25 (2019-01-15)
Fructosylation can significantly improve the solubility, stability and bioactivity of phenolic compounds, increasing their health benefits. Levansucrase from Gluconacetobacter diazotrophicus (LsdA, EC 2.4.1.10) was found to transfer the fructosyl unit of sucrose to different classes of phenolic compounds. Among the
M Ontiveros et al.
Biochemical pharmacology, 166, 1-11 (2019-05-10)
Research on flavonoids from plant sources has recently sparked increasing interest because of their beneficial health properties. Different studies have shown that flavonoids change the intracellular Ca2+ homeostasis linked to alterations in the function of mitochondria, Ca2+ channels and Ca2+

Articles

The C-terminal c-Src kinase (Csk) is a 50-kDa cytosolic tyrosine kinase expressed in all examined cell types

Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service