MilliporeSigma
Natural flavonoids inhibit the plasma membrane Ca2+-ATPase.
Biochemical pharmacology (2019-05-10)
M Ontiveros, D Rinaldi, M Marder, M V Espelt, I Mangialavori, M Vigil, J P Rossi, M Ferreira-Gomes
ABSTRACT
Research on flavonoids from plant sources has recently sparked increasing interest because of their beneficial health properties. Different studies have shown that flavonoids change the intracellular Ca2+ homeostasis linked to alterations in the function of mitochondria, Ca2+ channels and Ca2+ pumps. These findings hint at plasma membrane Ca2+-ATPase (PMCA) involvement, as it transports Ca2+ actively to the extracellular medium coupled to ATP hydrolysis, thus maintaining ion cellular homeostasis. The present study aims to investigate the effect of several natural flavonoids on PMCA both in isolated protein systems and in living cells, and to establish the relationship between flavonoid structure and inhibitory activity on PMCA. Our results show that natural flavonoids inhibited purified and membranous PMCA with different effectiveness: quercetin and gossypin were the most potent and their inhibition mechanisms seem to be different, as quercetin does not prevent ATP binding whereas gossypin does. Moreover, PMCA activity was inhibited in human embryonic kidney cells which transiently overexpress PMCA, suggesting that the effects observed on isolated systems could occur in a complex structure like a living cell. In conclusion, this work reveals a novel molecular mechanism through which flavonoids inhibit PMCA, which leads to Ca2+ homeostasis and signaling alterations in the cell.