Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0071

Sigma-Aldrich

Trapidil

≥98% (HPLC)

Synonym(s):

AR 12008, Avantrin, N,N-diethyl-5-methyl-[1,2,4]Triazolo[1,5-a]pyrimidin-7-amine, Rocornal, Trapymin, Trapymine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5
CAS Number:
Molecular Weight:
205.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: ≥15 mg/mL

storage temp.

room temp

SMILES string

CCN(CC)c1cc(C)nc2ncnn12

InChI

1S/C10H15N5/c1-4-14(5-2)9-6-8(3)13-10-11-7-12-15(9)10/h6-7H,4-5H2,1-3H3

InChI key

GSNOZLZNQMLSKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trapidil was used to study the mechanism of osteoclastogenesis and bone loss in mice.

Biochem/physiol Actions

Trapidil is an antiplatelet agent that acts in part as a phosphodiesterase inhibitor and as a competitive inhibitor of the platelet-derived growth factor (PDGF) receptor. Trapidil, with its vasodilator and NO releasing effect may have some potential to diminish the tissue injury. Trapidil suppresses platelet-derived growth factor (PDGF)-induced vascular smooth muscle cell (VSMC) proliferation by inhibiting Raf-1/extracellular signal-regulated kinase (ERK) via cAMP/protein kinase A (PKA). In addn. to cAMP/PKA activation, trapidil inhibits RhoA/ROCK activation.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaetano Ragno et al.
Analytical and bioanalytical chemistry, 389(3), 923-929 (2007-08-07)
A novel analytical technique able to determine the anti-ischemic drug trapidil in human serum and urine is proposed. In order to achieve satisfactory sensitivity and selectivity, an extraction procedure was required to isolate the drug from complex matrixes such as
Zhi-Wen Zhang et al.
Neurosurgery, 66(4), 728-735 (2010-03-23)
After subarachnoid hemorrhage (SAH), platelet-derived growth factor-BB (PDGF-BB) is secreted in and around the cerebral arteries. To clarify the role of PDGF-BB in the development of vasospasm after SAH, we determined whether PDGF-BB alone can cause long-lasting vasoconstriction of a
Hisao Ogawa et al.
Journal of cardiology, 54(2), 171-182 (2009-09-29)
"Evidence-based medicine (EBM)" implies effective and high quality practice for patients based on well-grounded medical science. The success of clinical trials in Japan is essential to build original evidence specific for Japanese patients. Based on this concept, we have performed
Russell T Turner et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 25(7), 1637-1649 (2010-03-05)
Chronic hyperparathyroidism (HPT) is a common cause of metabolic bone disease. These studies investigated the underlying cellular and molecular mechanisms responsible for the detrimental actions of elevated parathyroid hormone (PTH) on the skeleton. Bone biopsies from hyperparathyroid patients revealed an
Salih Somuncu et al.
International journal of urology : official journal of the Japanese Urological Association, 13(5), 601-605 (2006-06-15)
We aimed to detect the preventive effects of trapidil in ischemia-reperfusion (IR) injury due to testicular torsion and detorsion. Forty prepubertal albino rats were used. In the IR group, torsion was created by rotating the left testis over 2 h

Articles

Cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and/or cGMP. There are 11 different mammalian PDE families.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service