MilliporeSigma
  • Clinical evidence for Japanese population based on prospective studies--linking clinical trials and clinical practice.
Clinical evidence for Japanese population based on prospective studies--linking clinical trials and clinical practice.
Journal of cardiology (2009-09-29)
Hisao Ogawa, Sunao Kojima
ABSTRACT
"Evidence-based medicine (EBM)" implies effective and high quality practice for patients based on well-grounded medical science. The success of clinical trials in Japan is essential to build original evidence specific for Japanese patients. Based on this concept, we have performed several large-scale clinical trials to provide EBM, including the Japanese Antiplatelets Myocardial Infarction Study [JAMIS; clinical improvement in acute myocardial infarction (AMI) patients with antiplatelet therapy], the Japanese beta-Blockers and Calcium Antagonists Myocardial Infarction (JBCMI; comparison of the effects of beta-blockers and calcium antagonists on cardiovascular events in post-AMI patients), a multicenter study for aggressive lipid-lowering strategy by HMG-CoA reductase inhibitors in patients with AMI (MUSASHI; effects of statin therapy on cardiovascular events in patients with AMI), and the Japanese Primary Prevention of Atherosclerosis with Aspirin for Diabetes (JPAD trial; efficacy of low-dose aspirin therapy for primary prevention of atherosclerotic events in type 2 diabetic patients). The results of these prospective studies were directly linked with clinical practice. We have acquired the know-how of large-scale clinical trials; an important point is to have passion for "buildup evidence specific for the Japanese" and to recruit subjects for enrollment after explaining the significance of "clinical trials for the Japanese".
Product Description
Trapidil, European Pharmacopoeia (EP) Reference Standard