Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00919

Sigma-Aldrich

Cichoriin

≥95% (HPLC)

Synonym(s):

Cichoriin, 6,7-Dihydroxycoumarin 7-glucoside, 6-Hydroxy-7-(β-D-glucopyranosyloxy)coumarin, Cichorioside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O9
CAS Number:
Molecular Weight:
340.28
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

Assay

≥95% (HPLC)

form

powder

color

off-white to beige

mp

213—214 °C

storage temp.

2-8°C

InChI

1S/C15H16O9/c16-5-10-12(19)13(20)14(21)15(24-10)23-9-4-8-6(3-7(9)17)1-2-11(18)22-8/h1-4,10,12-17,19-21H,5H2/t10-,12-,13+,14-,15-/m1/s1

InChI key

WNBCMONIPIJTSB-TVKJYDDYSA-N

General description

Cichoriin, also known as cichorioside, is a coumarin glycoside. It is a bioactive natural compound commonly derived from plants such as Cichorium intybus, Cichorium endivia, Koelpinia linearis, Lactuca altaica, Chondrilla juncea, and Lactuca tatarica. Current research indicates that this metabolite may possess various biological activities, including antioxidant, neuroprotective, and antidiabetic properties.

Application

Cichoriin finds application in compound screening libraries, metabolomics, biochemical, phytochemical, and pharmaceutical research.

Features and Benefits

High quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Probing binding specificity of the sucrose transporter AtSUC2 with fluorescent coumarin glucosides.
De Moliner
Journal of Experimental Botany, 69, 2473-2482 (2018)
Chemical Composition and Nutritive Benefits of Chicory (Cichorium intybus) as an Ideal Complementary and/or Alternative Livestock Feed Supplement
Nwafor
TheScientificWorldJournal, 2017 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service