Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04480585

Luteolin-7-O-β-D-glucuronide

primary reference standard

Synonym(s):

2-(3,4-Dihydroxyphenyl)-5-hydroxy-4-oxo-4H-1-benzopyran-7-yl-β-D-glucopyranosiduronic acid, 3′,4′,5,7-Tetrahydroxy-7-β-D-glucopyranuronoside flavone, 3′,4′,5,7-Tetrahydroxyflavone 7-glucuronide, Cyanidenon 7-glucuronide, Cyanidenon-7-O-β-D-glucuronic acid, Luteolin 7-O-β-D-glucuronopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$629.00

$629.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$629.00

About This Item

Empirical Formula (Hill Notation):
C21H18O12
CAS Number:
Molecular Weight:
462.36
UNSPSC Code:
85151701
NACRES:
NA.24

$629.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)c(O)c4

InChI

1S/C21H18O12/c22-9-2-1-7(3-10(9)23)13-6-12(25)15-11(24)4-8(5-14(15)32-13)31-21-18(28)16(26)17(27)19(33-21)20(29)30/h1-6,16-19,21-24,26-28H,(H,29,30)/t16-,17-,18+,19-,21?/m0/s1

InChI key

VSUOKLTVXQRUSG-DAZJWRSOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service