Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00373

Sigma-Aldrich

N-Acetylglucosaminyl-β-1,2-mannose

≥94% (HPLC)

Synonym(s):

N-Acetylglucosaminyl-β-1,2-mannose, β-1-2 N-acetylglucosamine mannose, 2-O-(2-acetamido-2-deoxy-β-D-glucopyranosyl)-D-mannopyranose, GlcNAc-β-1,2-Man, N-Acetyl-glucosamine mannose, β-D-GlcNAc-(1→2)-D-Man

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$413.00

$413.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$413.00

About This Item

Empirical Formula (Hill Notation):
C14H25NO11
CAS Number:
Molecular Weight:
383.35
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

$413.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥94% (HPLC)

form

powder

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1O[C@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO)=O

InChI

1S/C14H25NO11/c1-5(19)15-9-13(24)12(23)8(4-18)26-14(9)25-7(3-17)11(22)10(21)6(20)2-16/h3,6-14,16,18,20-24H,2,4H2,1H3,(H,15,19)/t6-,7-,8-,9-,10-,11-,12-,13-,14-/m1/s1

InChI key

WCIQTQUMKQRLKV-ZTGOBPNGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

N-Acetyglucosaminyl-beta-1,2-mannose is a common oligosaccharide that inhibits the adherence of typical and atypical respiratory pathogens.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service