Skip to Content
MilliporeSigma
N-Acetylglucosaminyl-β-1,2-mannose ≥94% (HPLC)

N-Acetylglucosaminyl-β-1,2-mannose

N-Acetylglucosaminyl-β-1,2-mannose ≥94% (HPLC)
Synonym(s):
N-Acetylglucosaminyl-β-1,2-mannose, β-D-GlcNAc-(1→2)-D-Man
Empirical Formula (Hill Notation):
C14H25NO11
CAS Number:
Molecular Weight:
383.35

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing