Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SEP030

Millipore

Seppro® IgY14

LC5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

column D × L

12.7 mm × 39.5 mm

capacity

5 mg total protein loading(71 μL of human plasma, average human protein concentration 70 mg/mL)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Seppro® IgY14 columns allow users to separate 14 highly abundant proteins (HAP) from plasma protein mixtures. The Avian polyclonal IgY (Immunoglobulin Yolk) antibodies, have a distinct, highly specific partitioning advantage over competing products, allowing the researcher to look at a much cleaner human protein mixture. Seppro® columns make further proteomics analysis into the plasma proteome much easier and provide cleaner results.

Proteins Depleted:

Albumin
IgG
α1-Antitrypsin
IgA
IgM
Transferrin
Haptoglobin
α2-Macroglobulin
Fibrinogen
Complement C3
α1-Acid Glycoprotein (Orosomucoid)
HDL (Apolipoproteins A-I and A-II)
LDL (mainly Apolipoprotein B)

Application

Seppro® IgY14, LC5 (5 mL bed volume columns) is designed for use as a plasma/serum protein removal tool base upon immune affinity removal of 14 highly abundant proteins (HAP) from human plasma protein mixtures via liquid chromatography. Seppro® IgY14 is used to support the analysis of low abundance human serum/plasma proteins.

Legal Information

Seppro is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • S4199Seppro® Dilution bufferSDS

  • S4324Seppro® Stripping BufferSDS

  • S4449Seppro® Neutralization BufferSDS

  • CLS8163Corning® Costar® Spin-X® centrifuge tube filters, cellulose acetate membrane, pore size 0.45 μm, non-sterileSDS

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Conhaim et al.
American journal of respiratory and critical care medicine, 153(2), 677-683 (1996-02-01)
Lung liquid conductance (Kf) is calculated as the quotient of lung lymph flow divided by net filtration pressure (Pnf), where Pnf is the balance of osmotic and hydrostatic pressures in the lung microcirculation. In protein depletion, lymph flow rises with
Does plasma protein depletion increase lung liquid conductance?
P Dodek
American journal of respiratory and critical care medicine, 154(4 Pt 1), 1213-1215 (1996-10-01)
Renato Millioni et al.
PloS one, 6(5), e19603-e19603 (2011-05-17)
To date, the complexity of the plasma proteome exceeds the analytical capacity of conventional approaches to isolate lower abundance proteins that may prove to be informative biomarkers. Only complex multistep separation strategies have been able to detect a substantial number

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service