Skip to Content
MilliporeSigma
All Photos(4)

Documents

SAB5702251

Sigma-Aldrich

Anti-YKL-40/CHI3L1 Antibody, clone 7T1R3, Rabbit Monoclonal

Synonym(s):

CHI3L1,ASRT7,CGP-39,GP-39,GP39,HC-gp39,HCGP-3P,YKL-40,YKL40,YYL-40,hCGP-39

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

7T1R3, monoclonal

form

liquid

mol wt

30-43 kDa

species reactivity

mouse, mouse

concentration

1.00 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:1000

color

colorless

isotype

IgG

immunogen sequence

PGRFTKEAGTLAYYEICDFLRGATVHRILGQQVPYATKGNQWVGYDDQESVKSKVQYLKDRQLAGAMVWALDLDDFQGSFCGQDLRFPLTNAIKDALAAT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHI3L1(1116)

General description

Chitinases catalyze the hydrolysis of chitin, which is an abundant glycopolymer found in insect exoskeletons and fungal cell walls. The glycoside hydrolase 18 family of chitinases includes eight human family members. This gene encodes a glycoprotein member of the glycosyl hydrolase 18 family. The protein lacks chitinase activity and is secreted by activated macrophages, chondrocytes, neutrophils and synovial cells. The protein is thought to play a role in the process of inflammation and tissue remodeling.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 284-383 of human YKL-40/CHI3L1 (NP_001267.2).
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service