Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5702039

Sigma-Aldrich

Anti-IL10 Antibody, clone 6U0G7, Rabbit Monoclonal

Synonym(s):

CSIF, GVHDS, IL-10, IL10A, TGIF

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$533.00

$533.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$533.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$533.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

6U0G7, monoclonal

form

liquid

mol wt

18 kDa

species reactivity

rat, human, rat

concentration

1.60mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MHSSALLCCLVLLTGVRASPGQGTQSENSCTHFPGNLPNMLRDLRDAFSRVKTFFQMKDQLDNLLLKESLLEDFKGYLGCQALSEMIQFYLEEVMPQAEN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL10(3586)

General description

The protein encoded by this gene is a cytokine produced primarily by monocytes and to a lesser extent by lymphocytes. This cytokine has pleiotropic effects in immunoregulation and inflammation. It down-regulates the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. This cytokine can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice suggested the function of this cytokine as an essential immunoregulator in the intestinal tract. Mutations in this gene are associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis.[provided by RefSeq, May 2011]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human IL10 (P22301).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service