Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4301853

Sigma-Aldrich

Anti-PLAG1

affinity isolated antibody

Synonym(s):

SGPA, ZNF912

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$524.00

$524.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$524.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$524.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1: 20-100
western blot: 1:200-1:1000

accession no.

NP_001108106

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLAG1(5324)

Immunogen

Synthetic peptide of human PLAG1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.4 PBS, 0.05% NaN3, 40% Glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanbin Zheng et al.
Molecular cancer research : MCR, 18(3), 364-374 (2019-11-24)
We recently developed a novel computational algorithm that incorporates Bayesian methodology to identify rhabdomyosarcoma disease genes whose expression level correlates with copy-number variations, and we identified PLAG1 as a candidate oncogenic driver. Although PLAG1 has been shown to contribute to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service