Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200726

Sigma-Aldrich

Anti-Myosin (Smooth) antibody, Mouse monoclonal

clone hSM-V, purified from hybridoma cell culture

Synonym(s):

Anti-SM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

hSM-V, monoclonal

species reactivity

quail, rabbit, rat, human, sheep, bovine, mouse, porcine, canine

concentration

~1.0 mg/mL

technique(s)

immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: 10-20 μg/mL using methacarn -fixed paraffin-embedded human tonsil sections
immunoprecipitation (IP): suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYH11(4629)

General description

Anti-Myosin (Smooth) antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hSM-V hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a mouse immunized with human uterus smooth muscle extract. Myosin together with actin belong to a family of contractile proteins. Myosin contains two heavy chains (~200 kDa each) and four light chains (15-26 kDa). The Myosin molecules which consist of two major regions: tail (rod) and head, aggregate through the tail region into filaments and interact through the head region with actin and with ATP.

Immunogen

human uterus smooth muscle extract

Application

Anti-Myosin (Smooth) antibody, Mouse monoclonal has been used in various immunochemical assays, including immunohistochemistry, immunoblotting, immunofluorescence, immunoprecipitation and fluorescence-activated cell sorting (FACS) / flow cytometry.

Biochem/physiol Actions

Myosin is involved in a variety of cellular processes including secretion, cytoplasmic streaming, locomotion, phagocytosis and cytokinesis. This antibody can be a useful tool in the identification and localization of smooth muscle elements, in the diagnosis and classification of mesenchymal tumors and in myosin expression studies in normal and malignant cell proliferations.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Complete Characterization of Cardiac Myosin Heavy Chain (223 kDa) Enabled by Size-Exclusion Chromatography and Middle-down Mass Spectrometry
Jin Y, et al.
Analytical Chemistry, 89, 4922-4922 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service