Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABC615

Sigma-Aldrich

Anti-TNFα Receptor Antibody, clone 13F9.1

clone 13F9.1, from mouse

Synonym(s):

Tumor necrosis factor receptor superfamily member 1A, Tumor necrosis factor receptor 1, TNF-R1, Tumor necrosis factor receptor type I, TNF-RI, TNFR-I, p55, p60, CD120a, Tumor necrosis factor receptor superfamily member 1A, membrane form, Tumor necrosis f

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$467.00

$467.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$467.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$467.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13F9.1, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TNF(7124)

General description

Tumor necrosis factor receptor superfamily member 1A (UniProt P19438; also known as CD120a, p55, p60, TNF-R1, TNF-RI, TNFR-I, Tumor necrosis factor-alpha receptor, Tumor necrosis factor receptor type 1) is encoded by the TNFRSF1A (also known as FPF, MS5, TNFAR, TNFAR55, TNFAR60, TNFR1) gene (Gene ID 7132) in human. It functions as a receptor for TNFSF2/TNF-alpha and homotrimeric TNFSF1/lymphotoxin-alpha. Upon activation, TNFalpha receptor mediates apoptotic, anti-viral, and other cellular signaling events. Binding of TNF-alpha to the extracellular domain of TNF receptor leads to homotrimerization. Proteolytic cleavage results in the relese of Tumor necrosis factor-binding protein 1 (TBP1). Defects in TNF receptor cause tumor necrosis factor receptor-associated periodic syndrome (TRAPS), a hereditary periodic fever syndrome characterized by recurrent fever, abdominal pain, localized tender skin lesions and myalgia.

Specificity

Expected to react with spliced isoforms 1/FL-TNFR1, 2, 4/Delta6-TNFR1, and 5, but not isoform 3.

Immunogen

GST-tagged recombinant protein corresponding to the extracellular domain of human TNFα Receptor.

Application

Anti-TNF apha Receptor Antibody, clone 13F9.1 is a highly specific mouse monoclonal antibody, that targets TNFα Receptor & has been tested in Western Blotting, and Immunohistochemistry.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected TNFα Receptor in 10 µg of HepG2 and RAW264 cell lysate.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected TNFα Receptor in human colon tissue and human colon cancer cells.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected TNFα Receptor in 10 µg of Jurkat cell lysate.

Target description

~50/43 kDa observed. 50.5 kDa (isoform 1/FL-TNFR1 calculated), 38.7 kDa (isoform 2 calculated), 25.6 kDa (isoform 4/Delta6-TNFR1 calculated), 24.2 kDa (isoform 5 calculated).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Bai et al.
Molecular neurobiology, 56(1), 278-291 (2018-04-27)
Previous studies have shown that tumor necrosis factor alpha (TNFα) is significantly increased in complete Freund's adjuvant (CFA)-treated temporomandibular joint (TMJ) tissues. However, it is unclear whether TNFα in the trigeminal nociceptive system contributes to the development of TMJ pain.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service