Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB2104912

Sigma-Aldrich

Anti-STAT3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-APRF, Anti-FLJ20882, Anti-MGC16063

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

88 kDa

species reactivity

guinea pig, mouse, rabbit, horse, bovine, dog, human, rat, sheep

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAT3(6774)

Immunogen

Synthetic peptide directed towards the middle region of human STAT3

Biochem/physiol Actions

STAT3 is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act a

Sequence

Synthetic peptide located within the following region: SNTIDLPMSPRTLDSLMQFGNNGEGAEPSAGGQFESLTFDMELTSECATS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Vella et al.
Cancers, 12(3) (2020-03-11)
Shwachman-Diamond syndrome (SDS) is a rare inherited bone marrow failure syndrome, resulting in neutropenia and a risk of myeloid neoplasia. A mutation in a ribosome maturation factor accounts for almost all of the cases. Lymphoid involvement in SDS has not
Ekaterina A Polyakova et al.
Heliyon, 7(11), e08491-e08491 (2021-12-14)
Hyperleptinemia potentiates the effects of many atherogenic factors, such as inflammation, platelet aggregation, migration, hypertrophy, proliferation of vascular smooth muscle cells, and endothelial cell dysfunction. The present study analysed the effects of long-term hyperleptinemia in an in vivo myocardial ischemia-reperfusion
Xiaoying Zhao et al.
Biological research, 53(1), 28-28 (2020-07-06)
Kidney ischemia-reperfusion injury is a common pathophysiological phenomenon in the clinic. A large number of studies have found that the tyrosine protein kinase/signal transducer and activator of transcription (JAK/STAT) pathway is involved in the development of a variety of kidney

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service