MilliporeSigma
  • Hyperleptinemia results in systemic inflammation and the exacerbation of ischemia-reperfusion myocardial injury.
Hyperleptinemia results in systemic inflammation and the exacerbation of ischemia-reperfusion myocardial injury.
Heliyon (2021-12-14)
Ekaterina A Polyakova, Evgeny N Mikhaylov, Michael M Galagudza, Evgeny V Shlyakhto
ABSTRACT
Hyperleptinemia potentiates the effects of many atherogenic factors, such as inflammation, platelet aggregation, migration, hypertrophy, proliferation of vascular smooth muscle cells, and endothelial cell dysfunction. The present study analysed the effects of long-term hyperleptinemia in an in vivo myocardial ischemia-reperfusion model to demonstrate whether the in vivo deleterious effect also affects cardiac structure and function. Rats were subcutaneously administered leptin for 8 days to estimate the involvement of the JAK/STAT pathway. Data from 58 male Wistar rats were included in the final analysis. Myocardial infarction (MI) was modelled by the 30-minute ligation of the main left coronary artery followed by 120-minute reperfusion. Hemodynamic measurements, electrocardiography monitoring, echocardiography, myocardial infarct size and area at risk, blood biochemical parameters, leptin, IL-6, TNF-alpha, FGF-21, and cardiomyocyte morphology were measured. The expression of JAK2, p-JAK2, STAT3, p-STAT3 was assessed by Western Blot analysis. Statistical analyses were performed using IBM SPSS Statistics v.26. Eight-day hyperleptinemia in rats leads to an increase in blood pressure and heart rate, myocardial hypertrophy, impaired LV function, the frequency of ischemic arrhythmias, dyslipidemia, systemic inflammation, and the size of induced myocardial infarction. Significance: The blockade of the JAK/STAT signalling pathway effectively reverses the negative effects of leptin, including increased blood pressure and total cholesterol.
Sigma-Aldrich
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-JAK2 (Tyr1007/1008) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody