S5253
Stearoyl chloride
≥99%, liquid
Synonym(s):
Octadecanoyl chloride
About This Item
Recommended Products
Assay
≥99%
form
liquid
refractive index
n20/D 1.454 (lit.)
bp
174-178 °C/2 mmHg (lit.)
mp
21-22 °C (lit.)
density
0.897 g/mL at 25 °C (lit.)
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCCC(Cl)=O
InChI
1S/C18H35ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3
InChI key
WTBAHSZERDXKKZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
replaced by
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
329.0 °F - closed cup
Flash Point(C)
165 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service