Skip to Content
MilliporeSigma

1.06049

SAFC

Dichloromethane

EMPROVE® ESSENTIAL, Ph. Eur., BP, NF

Pharma Manufacturing

Synonym(s):

DCM, Methylene chloride, Methylene dichloride, Methylene chloride

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
$121.00
25 L
$783.00

$121.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2.5 L
$121.00
25 L
$783.00

About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-838-9

$121.00


Please contact Customer Service for Availability

Request a Bulk Order

Agency

BP
NF
Ph. Eur.

Quality Level

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

product line

EMPROVE® ESSENTIAL

Assay

≥99% (GC)

form

liquid

autoignition temp.

1223 °F
605 °C (DIN 51794)

expl. lim.

22 %

technique(s)

API processing | polymorph screening: suitable

refractive index

n20/D 1.424 (lit.)

pH

7 (20 °C)

bp

39.8-40 °C (lit.)

mp

-95 °C
−95 °C (lit.)

solubility

20 g/L

density

1.325 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharmaceutical

storage temp.

2-30°C

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Methanol is used as solvent in a variety of pharmaceutical production processes, including tablet coating, liposome production, microsphere, and nanoparticle production.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service