Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S180

Sigma-Aldrich

SB-206553 hydrochloride hydrate

≥98% (HPLC), solid

Synonym(s):

5-Methyl-1-(3-pyridylcarbamoyl)-1,2,3,5-tetrahydropyrrolo[2,3-f]indole hydrochloride hydrate, N-3-Pyridinyl-3,5-dihydro-5-methylbenzo(1,2-b:4,5-b′)dipyrrole-1(2H)carboxamide hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N4O · HCl · xH2O
CAS Number:
Molecular Weight:
328.80 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

yellow

solubility

H2O: >10 mg/mL

SMILES string

O.Cl.Cn1ccc2cc3N(CCc3cc12)C(=O)Nc4cccnc4

InChI

1S/C17H16N4O.ClH.H2O/c1-20-7-4-12-10-16-13(9-15(12)20)5-8-21(16)17(22)19-14-3-2-6-18-11-14;;/h2-4,6-7,9-11H,5,8H2,1H3,(H,19,22);1H;1H2

InChI key

NXGFRKQJHLVHIT-UHFFFAOYSA-N

Gene Information

Application

SB-206553 hydrochloride hydrate has been used as a serotonin type 2B/2C receptor (5HT2B/2CR) blocker to study its effects on sheep mitral valves.

Biochem/physiol Actions

SB-206553 is a serotonin type 2B/2C receptor (5HT2B/2CR) antagonist.

Legal Information

Manufactured and sold under exclusive license from GlaxoSmithKline Pharmaceuticals

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R T Jensen et al.
Pharmacological reviews, 60(1), 1-42 (2007-12-07)
The mammalian bombesin receptor family comprises three G protein-coupled heptahelical receptors: the neuromedin B (NMB) receptor (BB(1)), the gastrin-releasing peptide (GRP) receptor (BB(2)), and the orphan receptor bombesin receptor subtype 3 (BRS-3) (BB(3)). Each receptor is widely distributed, especially in
O Yoshie et al.
Journal of biochemistry, 100(3), 531-541 (1986-09-01)
Highly purified recombinant human tumor necrosis factor (TNF) (molecular mass determined as 17 kilodaltons (kDa) by sodium dodecyl sulfate-polyacrylamide gel electrophoresis and as 36 kDa by Sephadex G-100 gel chromatography) was labeled with 125I to a specific activity of 5
I T Forbes et al.
Journal of medicinal chemistry, 39(25), 4966-4977 (1996-12-06)
The synthesis and biological activity are reported for a series of analogues of the previously published indole urea 2 (SB-206553), designed to probe the 5-HT(2C) receptor binding site. Small molecule modeling studies have been used to define a region in
Michael Hendricks et al.
Neural development, 2, 6-6 (2007-03-16)
Electroporation is a technique for the introduction of nucleic acids and other macromolecules into cells. In chick embryos it has been a particularly powerful technique for the spatial and temporal control of gene expression in developmental studies. Electroporation methods have
G Griebel et al.
Neuropharmacology, 36(6), 793-802 (1997-06-01)
Although there is some evidence that compounds acting at 5-HT2 receptors show anxiolytic activity, little is known about the specific involvement of the different 5-HT2 receptor subtypes in the modulation of anxiety-related responses. In the present study, the behavioural effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service