Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

S1266

Sigma-Aldrich

Sucrose monodecanoate

Synonym(s):

Capric acid sucrose ester, Sucrose monocaprate, n-Decanoylsucrose

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$510.00

$510.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$510.00

About This Item

Empirical Formula (Hill Notation):
C22H40O12
CAS Number:
Molecular Weight:
496.55
Beilstein:
4924317
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

$510.00


Please contact Customer Service for Availability

Request a Bulk Order

description

non-ionic

Quality Level

mol wt

496.55 g/mol

CMC

2.5

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C22H40O12/c1-2-3-4-5-6-7-8-9-15(25)31-11-14-16(26)18(28)19(29)21(32-14)34-22(12-24)20(30)17(27)13(10-23)33-22/h13-14,16-21,23-24,26-30H,2-12H2,1H3/t13-,14-,16-,17-,18+,19-,20+,21-,22+/m1/s1

InChI key

STVGXWVWPOLILC-LUQRLMJOSA-N

Looking for similar products? Visit Product Comparison Guide

Quality

Mixture of esters

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Pranee Inprakhon et al.
Journal of biotechnology, 259, 182-190 (2017-07-29)
Sucrose monocaprate was synthesized by carrying out a lipase-catalyzed transesterification in a non-aqueous biphasic medium. Vinyl caprate was mechanically dispersed into a solution of sucrose in DMSO. The use of DMSO allowed increasing sucrose concentration up to 0.7M (in DMSO).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service