Skip to Content
MilliporeSigma
All Photos(1)

Documents

324374

Millipore

n-Dodecanoylsucrose

Non-ionic detergent useful for solubilization of membrane-bound proteins in their native state. Absorbance (10%, H2O, 260 nm):0.5.

Synonym(s):

n-Dodecanoylsucrose, n-Monododecanoate-α-D-glucopyranoside, β-D-Fructofuranosylsucrose Monolaurate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H44O12
CAS Number:
Molecular Weight:
524.60
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

CMC

300 μM

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C12H22O11.C12H24O2/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12;1-2-3-4-5-6-7-8-9-10-11-12(13)14/h4-11,13-20H,1-3H2;2-11H2,1H3,(H,13,14)/t4-,5-,6-,7-,8+,9-,10+,11-,12+;/m1./s1

InChI key

PVVVEHXCVQRLOC-AKSHDPDZSA-N

General description

Absorbance (10%, H2O, 260 nm): <0.5.
Non-ionic detergent useful for solubilization of membrane-bound proteins in their native state (absorbance: <0.5 at 260 nm, 10%, H2O; CMC: 300 µM).
Non-ionic detergent useful for solubilization of membrane-bound proteins in their native state.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Makino, H., et al. 1983. Agric. Biol. Chem.47, 319.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service