Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0745

Sigma-Aldrich

Human TNFRSF17 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available. Contact Technical Service

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 48 pg/mL
standard curve range: 0.049-12 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFRSF17(608)

General description

The antibody pair provided in this kit recognizes human Tumor Necrosis Factor Receptor Superfamily Member 17.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xun Ma et al.
Gene therapy, 26(5), 187-197 (2019-03-31)
Patients with immunoglobulin (Ig) light-chain (LC) diseases such as LC light-chain amyloidosis die with organ failure and need new therapies. We sought a model to test anti-LC siRNA delivery to human plasma cells, requiring circulating LC, in vivo indicators of
Javier Rodríguez-Carrio et al.
Frontiers in immunology, 9, 2351-2351 (2018-10-30)
Introduction: B-cell activation is pivotal in rheumatoid arthritis (RA) pathogenesis and represents a relevant therapeutic target. The main aim of this study was to characterize the profiles of B-cell factors and their decoy receptors in RA and evaluate their clinical
Vianihuini Figueroa-Vazquez et al.
Molecular cancer therapeutics, 20(2), 367-378 (2020-12-11)
Despite major treatment advances in recent years, patients with multiple myeloma inevitably relapse. The RNA polymerase II complex has been identified as a promising therapeutic target in both proliferating and dormant cancer cells. Alpha-amanitin, a toxin so far without clinical
Elisabeth Schuh et al.
Journal of immunology (Baltimore, Md. : 1950), 198(8), 3081-3088 (2017-03-12)
The BAFF-APRIL system is best known for its control of B cell homeostasis, and it is a target of therapeutic intervention in autoimmune diseases and lymphoma. By analyzing the expression of the three receptors of this system, B cell maturation
Liang Lin et al.
Leukemia, 35(3), 752-763 (2020-07-08)
Chimeric antigen receptor (CAR) T-cell therapy remains limited to select centers that can carefully monitor adverse events. To broaden use of CAR T cells in community clinics and in a frontline setting, we developed a novel CD8+ CAR T-cell product

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service