Yes, the 20X wash buffer with the product code RABWASH4 should be diluted with deionized or distilled water, similar to the other components of the kit.
Select a Size
$75.00
Select a Size
About This Item
$75.00
Recommended Products
shipped in
wet ice
storage temp.
−20°C
Other Notes
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
The user guide refers to the wash buffer 20 x, with the product code RABWASH4; however, it does not provide instructions regarding how to dilute it. Should it be diluted with distilled water like the other components of the kit?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service