Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0542

Sigma-Aldrich

Human IL13RA2 / Interleukin-13 Receptor Subunit Alpha-2 ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)
sample type urine

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.4 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL13RA2(3598)

General description

The antibody pair provided in this kit detects Human Interleukin-13 Receptor Subunit Alpha-2 in serum, plasma, cell culture supernatants, and urine.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonakshi Bhattacharjee et al.
Cell reports, 28(1), 231-244 (2019-07-04)
Helicobacter pylori chronically colonizes the stomach and is strongly associated with gastric cancer. Its concomitant occurrence with helminths such as schistosomes has been linked to reduced cancer incidence, presumably due to suppression of H. pylori-associated pro-inflammatory responses. However, experimental evidence in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service