Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0014

Sigma-Aldrich

Human Angiogenin ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.77

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 pg/mL
standard curve range: 1.64-400 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ANG(283)

General description

The Human Angiogenin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Angiogenin in serum, plasma, cell culture supernates and urine.

Immunogen

Recombinant Human Angiostatin

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Charytan et al.
International journal of cardiology, 176(1), 99-109 (2014-07-23)
Sudden cardiovascular death is increased in chronic kidney disease (CKD). Experimental CKD models suggest that angiogenesis and nitric oxide (NO) inhibitors induce myocardial fibrosis and microvascular dropout thereby facilitating arrhythmogenesis. We undertook this study to characterize associations of CKD with
Sujoy Dutta et al.
Molecular oncology, 8(3), 483-507 (2014-01-25)
Angiogenin (ANG), a 14-kDa pro-angiogenic secreted protein, has been shown to play a role in cell migration and tumor invasion, which involve proteolytic cleavage of plasminogen to generate plasmin. However, the mechanism by which ANG regulates plasmin formation and cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service