Skip to Content
MilliporeSigma
All Photos(1)

Documents

R8814N

Sigma-Aldrich

Rat Spinal Cord Neurons: RSpN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

rat spinal cord

Quality Level

packaging

pkg of 2,000,000 cells (R8814N-20)
pkg of 4,000,000 cells (R8814N-40)

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

morphology

Adherent. When cultured under the recommended conditions, cells arborize and form complex neurite network in one week.

technique(s)

cell culture | mammalian: suitable

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


Rat Spinal Cord Neurons (RSpN) are derived from spinal cords of normal embryonic rat (gestation day 15).

Cell Line Origin

Spinal Cord

Application

Basic research on neural signals, somatosensory and motor functions, spinal cord injuries, diseases and degeneration, Friedreich′s ataxia, ALS, spinal muscular atrophies, disease mechanisms, pathophysiologies, drug screening and development, toxicity testing, immunostaining, live cell imaging, co-culturing, electrophysiology.

Packaging

R8814N-20 2,000,000 cells in Serum-free Freezing Medium
R8814N-40 4,000,000 cells in Serum-free Freezing Medium

Components

Serum-free Freezing Medium

Quality

Each lot was tested for proper morphology, Stain positive for β III-Tubulin, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service