Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R8810N-10

Sigma-Aldrich

Rat Midbrain Neurons: RMbN

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$910.00

$910.00


Available to ship onApril 10, 2025Details



Select a Size

Change View
1 EACH
$910.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$910.00


Available to ship onApril 10, 2025Details


biological source

rat midbrain

Quality Level

packaging

pkg of 1,000,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

morphology

Adherent. When cultured under the recommended conditions, cells arborize and form complex neurite network in one week.

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


Rat Midbrain Neurons (RMbN) are derived from midbrains of normal embryonic rat (gestation day 15). >1,000,000 cells cryopreserved in Serum-free Freezing Medium.

Cell Line Origin

Midbrain

Application

Basic research on visual and auditory systems, motor control, alertness, sleep/wake, temperature regulation, motivation, habituation, Parkinson′s disease, ALS, supranuclear palsy, gait, muscle control, disease mechanisms, pathophysiologies, drug screening and development, toxicity testing, immunostaining, live cell imaging, co-culturing, electrophysiology.

Components

Serum-free Freezing Medium

Quality

Each lot was tested for proper morphology, Stain positive for β III-Tubulin, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service