Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R7260

Sigma-Aldrich

Xba I from Xanthomonas badrii

Restriction Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

grade

for molecular biology

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Specificity

Recognition sequence: 5′-T/CTAGA-3′
Cutting results: A 2-10-fold Xba I overdigestion of 1 μg Ad-2 DNA substrate results in 100% cutting
Heat inactivation: Up to 15 units of enzyme inactivated at 65 °C for 15 minutes.

Application

XbaI is a restriction enxyme that cleaves DNA at the recognition site 5′-T/CTAGA-3′ to generate DNA fragments with 5′-cohesive termini.

Other Notes

Supplied with 10x Restriction Enzyme Buffer SH (B3657).
Comment: Xba I will only partially cleave DNA isolated from E. coli strains that have the dam methylase (dam+ strains). May cleave at secondary recognition sites under non-optimal conditions.

Physical form

Solution in 20 mM Tris-HCl, pH 8.0, 1 mM EDTA, 100 mM NaCl, 0.1 mM PMSF, 10 mM 2-mercaptoethanol, 50% glycerol (v/v), 0.02% polydocanol at 4 °C

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie L H Miller et al.
The Journal of biological chemistry, 279(32), 33528-33537 (2004-05-25)
The ubiquitin-interacting motif (UIM) is a short peptide motif with the dual function of binding ubiquitin and promoting ubiquitylation. This motif is conserved throughout eukaryotes and is present in numerous proteins involved in a wide variety of cellular processes including
A new specific endonuclease from Xanthomonas badrii..
Zain, B.S., and Roberts, R.J.,
Journal of Molecular Biology, 115, 244-255 (1977)
Miyuki Watabe et al.
The Ulster medical journal, 77(3), 168-174 (2008-10-30)
In Northern Ireland over the last 7 years, there is a mean of 41.9 laboratory reports per annum of human gastrointestinal infection (range 19-54) caused by Escherichia coli O157:H7. In the preceding years 1992-1996, reports were 5.4 per annum, whereas
E Malyguine et al.
Gene, 8(2), 163-177 (1980-01-01)
The specificity of XbaI, SalI, HhaI, PstI, BamHI and SstI is relaxed in the presence of an organic solvent, such as 20% glycerol or 8% dimethylsulfoxide (DMSO). This alteration, very pronounced in some cases, requires an excess of enzyme, varies
Jaroslav Jelinek et al.
Epigenetics, 7(12), 1368-1378 (2012-10-19)
Genome wide analysis of DNA methylation provides important information in a variety of diseases, including cancer. Here, we describe a simple method, Digital Restriction Enzyme Analysis of Methylation (DREAM), based on next generation sequencing analysis of methylation-specific signatures created by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service