Skip to Content
MilliporeSigma
All Photos(1)

Documents

PZ0417

Sigma-Aldrich

PF-06752469

≥98% (HPLC)

Synonym(s):

6-((3S,4S)-3,4-Dihydroxypyrrolidin-1-yl)-2-((S)-3-hydroxy-3-methylpyrrolidin-1-yl)-4-(trifluoromethyl)nicotinonitrile, 6-[(3S,4S)-3,4-Dihydroxy-1-pyrrolidinyl]-2-[(3S)-3-hydroxy-3-methyl-1-pyrrolidinyl]-4-(trifluoromethyl)-3-pyridinecarbonitrile, KHK-IN-2, PF 06752469, PF 675, PF 6752469, PF-675, PF-6752469, PF06752469, PF675, PF6752469

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19F3N4O3
CAS Number:
Molecular Weight:
372.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

PF-06752469 (PF-6752469) is an orally active, non-cytotoxic (300 μM, HepG2), potent and selective ketohexokinase (KHK) inhibitor (human KHK-A/C IC50 = 390/450 nM; rat KHK IC50 = 340 nM) with no significant activity against a broad panel of transporters, receptors, ion channels, and enzymes, including hERG and human CYP450s. PF-06752469 prevents fructose administration-induced fructose 1-phosphate (F1P) upregulation (hepatic/renal ED50 = 15.7/30.0 mg/kg p.o. 2 min prior to 500 mg fructose/10 mL/kg i.v. in rats) by inhibiting KHK-mediated fructose metabolism in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service