Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00285

Sigma-Aldrich

Glucose-Udp-Fluorescein Conjugate

≥95% (HPLC)

Synonym(s):

Glucose-UDP-Fluorescein Conjugate, Glucose-Uridine Diphosphate, Glucose-Uridine Diphosphate-(PEG)6-Fluorescein Conjugate, UDP-Glc-(PEG)6-Fluorescein Conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$688.00

$688.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$688.00

About This Item

UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.28

$688.00


Please contact Customer Service for Availability

Request a Bulk Order

conjugate

fluorescein conjugate

Assay

≥95% (HPLC)

form

solid

color

orange-brown, solid

fluorescence

λex 490 nm; λem 514 nm

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](OP(OP([O-])(OC[C@H]2O[C@@H](N(C=C3/C=C/CNC(C4=CC=C(C([O-])=O)C(C(C5=C(O6)C=C(O)C=C5)=C7C6=CC(C=C7)=O)=C4)=O)C(NC3=O)=O)[C@H](O)[C@@H]2O)=O)([O-])=O)O[C@H](CO)[C@H]1O.CC[NH+](CC)CC.CC[NH+](CC)CC.CC[NH+](CC)CC

InChI

1S/C39H39N3O23P2.3C6H15N/c43-14-26-29(46)31(48)33(50)38(63-26)64-67(58,59)65-66(56,57)60-15-27-30(47)32(49)36(62-27)42-13-17(35(52)41-39(42)55)2-1-9-40-34(51)16-3-6-20(37(53)54)23(10-16)28-21-7-4-18(44)11-24(21)61-25-12-19(45)5-8-22(25)28;3*1-4-7(5-2)6-3/h1-8,10-13,26-27,29-33,36,38,43-44,46-50H,9,14-15H2,(H,40,51)(H,53,54)(H,56,57)(H,58,59)(H,41,52,55);3*4-6H2,1-3H3/b2-1+;;;/t26?,27-,29-,30-,31-,32-,33?,36-,38+;;;/m1.../s1

InChI key

ZANXKXSOYBZRQT-VBPZYQLFSA-N

Biochem/physiol Actions

Glucose-UDP, or UDP-Glucose, is a nucleotide sugar that plays a key role in carbohydrate metabolism. UDP-Glucose is a precursor to glycogen and a key substrate for glycosyltransferases, such as Toxins A and B from Clostridium difficile. Fluorescent analogs of UDP-Glucose can be used for studying glucosyltransferases and clostridial toxins. This fluorescein UDP-Glucose derivative has spectral properties identical to those of FITC conjugates. Compared to SMB00284, this UDP-Glucose derivative has a much shorter spacer between the fluorophore and UDP moiety.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philippe Jean et al.
Nature communications, 11(1), 3208-3208 (2020-06-27)
Inner hair cells (IHCs) are the primary receptors for hearing. They are housed in the cochlea and convey sound information to the brain via synapses with the auditory nerve. IHCs have been thought to be electrically and metabolically independent from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service