Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0379

Sigma-Aldrich

PF-04827736

≥98% (HPLC)

Synonym(s):

(S)-7,8-Dimethoxy-N-[1-methyl-2-(5-methyl-1H-pyrazol-3-yl)ethyl]-4-quinazolinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N5O2
CAS Number:
Molecular Weight:
327.38
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

CC1=NNC(C[C@H](C)NC2=NC=NC3=C2C=CC(OC)=C3OC)=C1

Biochem/physiol Actions

PF-04827736 is a non-brain penetrant potent and selective inhibitor of phosphodiesterase PDE1, thought to be involved in regulation of vascular smooth muscle.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service