Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0119

Sigma-Aldrich

Rabbit anti-NONO Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

52 kDa subunit, 54 kDa nuclear RNA- and DNA-binding protein, 54-kD, 55 kDa nuclear protein, DNA-binding p52/p100 complex, MRXS34, NMT55, NRB54, NonO protein, Nuclear RNA-binding protein, P54, P54NRB, PPP1R114, non-Pou domain-containing octamer (ATGCAAAT) binding protein, p54(nrb), protein phosphatase 1, regulatory subunit 114

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
species reactivity:
rat, mouse, human
application:
IHC
IP
WB
technique(s):
immunohistochemistry: 1:200-1:1,000
immunoprecipitation (IP): 1- 4 μg/mg
western blot: 1:2,000- 1:10,000
citations:
1

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: 1:200-1:1,000
immunoprecipitation (IP): 1- 4 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_031389.3

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... NONO(4841)

Immunogen

The epitope recognized by PLA0119 maps to a region between residues 350 and 400 of human Non-POU Domain Containing, Octamer-binding using the numbering given in entry NP_031389.3 (GeneID 4841).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Non-POU-domain-containing octamer binding protein (NONO) is a member of the DBHS (drosophila behavior, human splicing) domain-containing family and is an RNA- and DNA- binding protein. NONO and other DBHS domain-containing proteins are multifunctional and are reported to be involved in transcriptional regulation, mRNA processing, and DNA non-homologous end joining (NHEJ). NONO functions as a coregulator of the androgen receptor (AR) and also regulates cAMP transcriptional activity by interacting with gene promoter elements. NONO is also involved in pre-mRNA splicing through an interaction with U5 snRNA and can stimulate DNA nonhomologous end joining (NHEJ) through the interaction with ku70/G22p and ku80/XRCC5 dimers.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service