Skip to Content
MilliporeSigma
All Photos(1)

Documents

P8800

Sigma-Aldrich

Heptaethylene glycol monododecyl ether

Synonym(s):

C12E7, Dodecylheptaglycol, Polyoxyethylene (7) lauryl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11(OCH2CH2)7OH
CAS Number:
Molecular Weight:
494.70
Beilstein:
1916066
EC Number:
MDL number:
UNSPSC Code:
12161900

description

non-ionic

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C26H54O8/c1-2-3-4-5-6-7-8-9-10-11-13-28-15-17-30-19-21-32-23-25-34-26-24-33-22-20-31-18-16-29-14-12-27/h27H,2-26H2,1H3

InChI key

DWHIUNMOTRUVPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service