Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8498

Sigma-Aldrich

Monoclonal Anti-Profilin antibody produced in mouse

~2 mg/mL, clone 2-11H7 (MAbPRF12a), purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2-11H7 (MAbPRF12a), monoclonal

form

buffered aqueous solution

mol wt

antigen ~14 kDa

species reactivity

Arabidopsis thaliana

concentration

~2 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: 1-2 μg/mL using extract of Arabidopsis sprout

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Arabidopsis thaliana ... PFN2(829056) , PRF1(816495) , PRF3(835761)

Looking for similar products? Visit Product Comparison Guide

General description

Profilins are highly conserved pan-allergens that occur in plant foods, pollen and latex. Profilins have five distinct isoforms (PRF1-5) that bind with actin filaments to regulate cytoskeletal organization. PRF1, 2 and 3 are constitutive profilins that occur in all vegetative cells. During the early stages of microsporogenesis, these isoforms are also expressed in microspores and ovules . Monoclonal Anti-Profilin antibody is specific for Arabidopsis constitutive profilin (PRF1 and PRF2 and weakly with PRF3). The antibody also reacts with tobacco profilin (approx. 14 kDa).

Immunogen

PRF1 protein.

Application

Monoclonal Anti-Profilin antibody is suitable for use in immunohistochemistry, western blot (1-2 μg/mL using extract of Arabidopsis sprout) and indirect ELISA.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kandasamy, M.K., et al.
Cell Motility, 52, 115-126 (2002)
Plant profilin isovariants are distinctly regulated in vegetative and reproductive tissues
Kandasamy, M., K., et al.
Cell Motility, 52, 22-32 (2002)
Alexandra Santos et al.
International archives of allergy and immunology, 155(3), 191-204 (2011-02-05)
Profilins are ubiquitous proteins, present in all eukaryotic cells and identified as allergens in pollen, latex and plant foods. The highly conserved structure justifies the cross-reactive nature of IgE antibodies against plant profilins and their designation as pan-allergens. Primary sensitization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service