Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7209

Sigma-Aldrich

1-Pyrenedodecanoic acid

≥90% (TLC)

Synonym(s):

12-(1-Pyrenyl)dodecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32O2
CAS Number:
Molecular Weight:
400.55
Beilstein:
2481235
MDL number:
UNSPSC Code:
12352200

Assay

≥90% (TLC)

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCCCCCCCc1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C28H32O2/c29-26(30)14-9-7-5-3-1-2-4-6-8-11-21-15-16-24-18-17-22-12-10-13-23-19-20-25(21)28(24)27(22)23/h10,12-13,15-20H,1-9,11,14H2,(H,29,30)

InChI key

JORFLUCVQONOPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Probe for studying uptake into cultured cells by FCM

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Viani et al.
Biochimica et biophysica acta, 1064(1), 24-30 (1991-04-26)
Two different pyrene derivatives, namely 12-(1-pyrene)dodecanoic acid (P12-FA) and N-(12-(1-pyrene)dodecanoyl)-galactosylsphingosine I3-sulfate (P12-CS) have been used to follow lipid peroxidation both in model and natural membranes. The malondialdehyde (MDA) production in small unilamellar vesicles of dipalmitoylphosphatidylcholine/arachidonic acid (80:20, molar ratio), symmetrically
K Fukuzawa et al.
BioFactors (Oxford, England), 7(1-2), 31-40 (1998-04-02)
The rate constants (ks) of 1O2 scavenging for alpha-tocopherol (alpha-Toc) and beta-carotene (beta-Car) were measured in liposome membranes, and compared with those in EtOH solution. 1O2 was site-specifically generated by photoirradiation using two photosensitizers, water-soluble Rose bengal (RB) and lipid-soluble
G Celedón et al.
The American journal of physiology, 275(6 Pt 1), C1429-C1431 (1998-12-09)
Because it has been reported that hypoxia in rats may promote lipid peroxidation and other free radical reactions that could modify membrane lipids and proteins, the effect of acute hypobaric hypoxia on human erythrocyte membranes was investigated. 12-(1-Pyrene)dodecanoic acid fluorescent
S Gatt et al.
The Biochemical journal, 253(2), 377-380 (1988-07-15)
Aqueous dispersions of 12-(1-pyrene)-dodecanoic acid (P12), a medium-chain fatty acid to which the fluorescent probe pyrene has been covalently linked, shows a considerable increase in fluorescence when the probe is introduced into a hydrophobic environment. This enables the uptake of
S Oshima et al.
Journal of nutritional science and vitaminology, 39(6), 607-615 (1993-12-01)
Large unilamellar liposomes comprising of egg yolk phosphatidylcholine (PC) was exposed to photoirradiation in the presence of methylene blue (water-soluble photosensitizer) or 12-(1-pyrene)dodecanoic acid (P-12, lipid-soluble photosensitizer) to estimate the inhibitory effect of beta-carotene and astaxanthin on photosensitized oxidation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service