MilliporeSigma
  • Singlet oxygen scavenging by alpha-tocopherol and beta-carotene: kinetic studies in phospholipid membranes and ethanol solution.
Singlet oxygen scavenging by alpha-tocopherol and beta-carotene: kinetic studies in phospholipid membranes and ethanol solution.
BioFactors (Oxford, England) (1998-04-02)
K Fukuzawa, Y Inokami, A Tokumura, J Terao, A Suzuki
ABSTRACT
The rate constants (ks) of 1O2 scavenging for alpha-tocopherol (alpha-Toc) and beta-carotene (beta-Car) were measured in liposome membranes, and compared with those in EtOH solution. 1O2 was site-specifically generated by photoirradiation using two photosensitizers, water-soluble Rose bengal (RB) and lipid-soluble 12-(1-pyrene)-dodecanoic acid (PDA). The ks value for beta-Car in EtOH solution was 1.3 x 10(10) M-1 s-1, which was 36 times that for alpha-Toc (3.6 x 10(8) M-1 s-1), but there was no difference between their ks values in liposomes (1.8 x 10(7) M-1 s-1 for beta-Car and 1.2 x 10(7) M-1 s-1 for alpha-Toc). In the liposomes, the ks value for alpha-Toc was affected by the membrane site where 1O2 was generated, which depended on the localization of the photosensitizer, being high at the membrane surface in the RB-system and low in the inner region of the membrane in the PDA-system. In contrast, the ks value for beta-Car was not affected by the 1O2-generating site. These differences were supposed to be caused by differences in the relative concentrations of 1O2 and active sites of alpha-Toc and beta-Car in the membranes. alpha-Toc and beta-Car inhibited 1O2-dependent peroxidation of egg yolk phosphatidylcholine (egg PC). The concentrations of alpha-Toc required for 50% inhibition of lipid peroxidation (IC50) were higher than those of beta-Car, being more than 6 times higher in EtOH solution and less than 2 times higher in liposomes. The ratio of the antioxidant activity of beta-Car to that of alpha-Toc was more in EtOH solution than in liposomes, and was well correlated with the ratio of their 1O2 scavenging rate constants.