Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5988

Sigma-Aldrich

PA28 Activator from rabbit

≥95% (SDS-PAGE), buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

Quality Level

Assay

≥95% (SDS-PAGE)

form

buffered aqueous solution

mol wt

170 kDa

shipped in

dry ice

storage temp.

−70°C

General description

Manufactured for Sigma by Boston Biochem., Inc.

Biochem/physiol Actions

Protein activator of the 20S proteasome. Enhances the 20S proteasome hydrolysis of peptide substrates, but not hydrolysis of ubiquitinated proteins.

Physical form

Solution in 50 mM Hepes, pH 7.6, 250 mM NaCl, 1 mM EDTA, and 1 mM DTT.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Dubiel et al.
The Journal of biological chemistry, 267(31), 22369-22377 (1992-11-05)
We have identified and purified a protein complex from human red blood cells that activates the multicatalytic protease (MCP). The complex, which we call the regulator, sediments at 11 S and is composed of 30-kDa subunits. The regulator does not
M Chu-Ping et al.
Biochimica et biophysica acta, 1119(3), 303-311 (1992-03-12)
An inhibitory protein for the 20S proteasome (also known as macropain, the multicatalytic proteinase complex and 20S proteinase) has been purified from bovine red blood cells. The inhibitor has an apparent molecular weight of 31,000 on SDS-PAGE and appears to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service