Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4502104

Sigma-Aldrich

Anti-ZDHHC9 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

DHHC-9, DHHC9, Palmitoyltransferase ZDHHC9, Zinc finger DHHC domain-containing protein 9, Zinc finger protein 379

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$698.00

$698.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$698.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$698.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZDHHC9(51114)

General description

Anti-ZDHHC9 Antibody detects endogenous levels of total ZDHHC9 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human ZDHHC9.

Immunogen Range: 315-364

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activity-dependent post-translational regulation of palmitoylating and depalmitoylating enzymes in the hippocampus.
Abazari, et al.
Journal of Cell Science, 136 (2023)
Francisco S Mesquita et al.
Developmental cell, 56(20), 2790-2807 (2021-10-03)
SARS-CoV-2 virions are surrounded by a lipid bilayer that contains membrane proteins such as spike, responsible for target-cell binding and virus fusion. We found that during SARS-CoV-2 infection, spike becomes lipid modified, through the sequential action of the S-acyltransferases ZDHHC20

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service