Skip to Content
MilliporeSigma
All Photos(1)

Documents

P5624

Sigma-Aldrich

PD-118057

≥98% (HPLC), solid

Synonym(s):

2-[[4-[2-(3,4-Dichlorophenyl)ethyl]phenyl]amino]benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17Cl2NO2
CAS Number:
Molecular Weight:
386.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

light yellow

solubility

DMSO: 9 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)c1ccccc1Nc2ccc(CCc3ccc(Cl)c(Cl)c3)cc2

InChI

1S/C21H17Cl2NO2/c22-18-12-9-15(13-19(18)23)6-5-14-7-10-16(11-8-14)24-20-4-2-1-3-17(20)21(25)26/h1-4,7-13,24H,5-6H2,(H,25,26)

InChI key

ZCQOSCDABPVAFB-UHFFFAOYSA-N

Biochem/physiol Actions

PD-118057 is an activator of ether-a-go-go-related (hERG) potassium channel. PD-118057 is a second identified hERG channel activator, a representative in the series of structural analogs; PD-118057 at 10 μM leads to a 111% current increase in rabbit ventricular wedge (in comparison RPR260243 leads to only a 15% increase at the same concentration);PD-118057, unlike RPR260243, does not have a major effect on gating or kinetic properties of this channel. PD-118057prevents and reverses QT interval prolongation; Compounds such as PD-118057 may offer new approach in the treatment of delayed repolarization conditions, which occur in inherited or acquired long QT syndrome and congestive heart failure

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to potassium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service