Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5544

Sigma-Aldrich

(R)-(+)-Propranolol hydrochloride

Synonym(s):

(R)-1-(isopropylamino)-3-(1-naphthyloxy)-2-propanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCH2CH(OH)CH2NHCH(CH3)2·HCl
CAS Number:
Molecular Weight:
295.80
Beilstein:
5780490
EC Number:
MDL number:
UNSPSC Code:
12352103

optical activity

[α]21/D +22.2°, c = 1.0 in ethanol(lit.)

mp

196-198 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl.CC(C)NC[C@@H](O)COc1cccc2ccccc12

InChI

1S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18H,10-11H2,1-2H3;1H/t14-;/m1./s1

InChI key

ZMRUPTIKESYGQW-PFEQFJNWSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Less active enantiomer of propranolol.

Caution

Shelf-life of the solid is at least four years if stored protected from the uptake of moisture.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service