Skip to Content
MilliporeSigma
(R)-(+)-Propranolol hydrochloride ≥98% (TLC)

(R)-(+)-Propranolol hydrochloride

(R)-(+)-Propranolol hydrochloride ≥98% (TLC)
Synonym(s):
(R)-1-(isopropylamino)-3-(1-naphthyloxy)-2-propanol hydrochloride
Linear Formula:
C10H7OCH2CH(OH)CH2NHCH(CH3)2·HCl
CAS Number:
Molecular Weight:
295.80
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing