Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5454

Sigma-Aldrich

Protein Kinase Cβ1 Peptide

lyophilized powder

Synonym(s):

Cys[ACM]-Ala-Gly-Phe-Ser-Tyr-Thr-Asn-Pro-Glu-Phe-Val-Ile-Asn-Val, PKC β1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

form

lyophilized powder

Quality Level

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(NC(=O)C(Cc1ccccc1)NC(=O)C(CCC(O)=O)NC(=O)C2CCCN2C(=O)C(CC(N)=O)NC(=O)C(NC(=O)C(Cc3ccc(O)cc3)NC(=O)C(CO)NC(=O)C(Cc4ccccc4)NC(=O)CNC(=O)C(C)NC(=O)C(N)CSCNC(C)=O)C(C)O)C(C)C)C(=O)NC(CC(N)=O)C(=O)NC(C(C)C)C(O)=O

InChI

1S/C79H114N18O24S/c1-10-41(6)64(76(117)90-54(33-58(81)102)72(113)94-63(40(4)5)79(120)121)95-75(116)62(39(2)3)93-70(111)52(31-46-20-15-12-16-21-46)88-68(109)50(27-28-61(105)106)87-74(115)57-22-17-29-97(57)78(119)55(34-59(82)103)91-77(118)65(43(8)99)96-71(112)53(32-47-23-25-48(101)26-24-47)89-73(114)56(36-98)92-69(110)51(30-45-18-13-11-14-19-45)86-60(104)35-83-66(107)42(7)85-67(108)49(80)37-122-38-84-44(9)100/h11-16,18-21,23-26,39-43,49-57,62-65,98-99,101H,10,17,22,27-38,80H2,1-9H3,(H2,81,102)(H2,82,103)(H,83,107)(H,84,100)(H,85,108)(H,86,104)(H,87,115)(H,88,109)(H,89,114)(H,90,117)(H,91,118)(H,92,110)(H,93,111)(H,94,113)(H,95,116)(H,96,112)(H,105,106)(H,120,121)

InChI key

DOFZBSAZZNPGLN-UHFFFAOYSA-N

Specificity

Synthetic peptide corresponding to the C-terminal region of PKC β1 (658-671).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service